2(5H)-Furanone ≥96%

Supplier: Matrix Scientific

Synonyms: Furan-2(5H)-one

070417-25G 070417-5G
101944-654EA 53.85 USD
101944-654 101944-656
2(5H)-Furanone ≥96%
2(5H)-Furanone

Hazard code(s): Irritant

Formula: C₄H₄O₂
MW: 84.07 g/mol
Boiling Pt: 212…214 °C
Melting Pt: 3…4 °C
Density: 1.197 g/cm³
MDL Number: MFCD00005376
CAS Number: 497-23-4
EINECS: 207-839-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR