4-Amino-3-methylpyridine ≥98%

Synonyms: 4-Amino-3-picoline3-Methylpyridin-4-amine3-Methyl-4-pyridylamine

070055-5G
101943-114EA 253.52 USD
101943-114
4-Amino-3-methylpyridine ≥98%
4-Amino-3-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₈N₂
MW: 108.14 g/mol
Melting Pt: 106…107 °C
MDL Number: MFCD01704431
CAS Number: 1990-90-5
EINECS: 217-872-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR