4-Bromoquinoline ≥98%

Supplier: Matrix Scientific
Danger

069910-1G
101942-502EA 315.38 USD
101942-502
4-Bromoquinoline ≥98%
4-Bromoquinoline

Hazard code(s): Irritant

Formula: C₉H₆BrN
MW: 208.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07644514
CAS Number: 3964-04-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR