6-Isoquinolinol ≥97%

Supplier: Matrix Scientific

Synonyms: 6-HydroxyisoquinolineIsoquinolin-6-ol

069893-1G
101942-426EA 44.85 USD
101942-426
6-Isoquinolinol ≥97%
6-Isoquinolinol

Hazard code(s): Irritant

Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD04114860
CAS Number: 7651-82-3
EINECS: 231-612-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR