2-Formyl-3-thiopheneboronic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Formylthiophene-3-boronic acid
, (2-Formylthiophen-3-yl)boronic acid
069830-1G
101942-122EA
233.85
USD
101942-122
2-Formyl-3-thiopheneboronic acid ≥95%
2-Formyl-3-thiopheneboronic acid
Hazard code(s): Irritant
Formula:
C₅H₅BO₃S MW: 155.97 g/mol |
MDL Number:
MFCD01075678 CAS Number: 4347-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...