5-(Trimethylsilyl)thiazole ≥97%

Supplier: Matrix Scientific

069804-5G
101942-010EA 557.75 USD
101942-010
5-(Trimethylsilyl)thiazole ≥97%
5-(Trimethylsilyl)thiazole

Hazard code(s): Irritant

Formula: C₆H₁₁NSSi
MW: 157.31 g/mol
MDL Number: MFCD00191506
CAS Number: 79265-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR