1-Boc-4-methylindole-2-boronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: N-(tert-Butoxycarbonyl)-4-methylindole-2-boronic acid

069619-5G 069619-1G
101941-118EA 1073.85 USD
101941-118 101941-116
1-Boc-4-methylindole-2-boronic acid ≥97%
1-Boc-4-methylindole-2-boronic acid

Hazard code(s): Irritant

Formula: C₁₄H₁₈BNO₄
MDL Number: MFCD09265525
CAS Number: 352359-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR