1-Boc-4-chloroindole-2-boronic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
(1-(tert-Butoxycarbonyl)-4-chloro-1H-indol-2-yl)boronic acid
, 4-Chloro-N-(Boc)-indole-2-boronic acid
069518-25G
069518-1G
069518-5G
101940-572EA
263.08
USD
101940-572
101940-574
101940-576
1-Boc-4-chloroindole-2-boronic acid ≥98%
1-Boc-4-chloroindole-2-boronic acid
Hazard code(s): Irritant
Formula:
C₁₃H₁₅BClNO₄ MW: 295.53 g/mol |
MDL Number:
MFCD04114582 CAS Number: 475102-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...