2-Fluoro-4-pyridineboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (2-Fluoropyridin-4-yl)boronic acid2-Fluoropyridine-4-boronic acid2-Fluoro-4-pyridinylboronic acid

069499-5G 069499-1G 069499-25G
101940-472EA 976.92 USD
101940-472 101940-470 101940-468
2-Fluoro-4-pyridineboronic acid ≥97%
2-Fluoro-4-pyridineboronic acid

Hazard code(s): Irritant

Formula: C₅H₅BFNO₂
MW: 140.91 g/mol
MDL Number: MFCD04112534
CAS Number: 401815-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR