1-Boc-5-fluoroindole-2-boronic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Boc-5-fluoroindole-2-boronic Acid
, N-(Boc)-5-fluoroindole-2-boronic acid
069487-5G
069487-1G
069487-25G
101940-402EA
1410.11
USD
101940-402
101940-404
101940-400
1-Boc-5-fluoroindole-2-boronic acid ≥95%
1-Boc-5-fluoroindole-2-boronic acid
Hazard code(s): Irritant, Refrigerate
Formula:
C₁₃H₁₅BFNO₄ MW: 279.07 g/mol |
MDL Number:
MFCD04114581 CAS Number: 352359-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...