4-Bromo-2-methylimidazole ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-methyl-1H-imidazole

069438-25G
101940-124EA 250.77 USD
101940-124
4-Bromo-2-methylimidazole ≥98%
4-Bromo-2-methylimidazole

Hazard code(s): Irritant

Formula: C₄H₅BrN₂
MW: 161 g/mol
MDL Number: MFCD03411959
CAS Number: 16265-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR