3-Chloro-1-iodopropane ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: 1-Chloro-3-iodopropane

069375-25G
101939-842EA 104.62 USD
101939-842
3-Chloro-1-iodopropane ≥95%
3-Chloro-1-iodopropane

Hazard code(s): Irritant

Formula: C₃H₆ClI
MW: 204.44 g/mol
Boiling Pt: 171 °C (1013 hPa)
Density: 1.904 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001096
CAS Number: 6940-76-7
EINECS: 230-088-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR