2-Bromo-4-chloropyridine ≥95%
Supplier: Matrix Scientific
069222-25G
069222-5G
101939-346EA
340
USD
101939-346
101939-344
2-Bromo-4-chloropyridine ≥95%
2-Bromo-4-chloropyridine
Hazard code(s): Irritant
Formula:
C₅H₃BrClN |
MDL Number:
MFCD03840757 CAS Number: 22918-01-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...