3-Amino-6-methylpyridazine ≥95%

Supplier: Matrix Scientific

068663-5G
101937-146EA 730.77 USD
101937-146
3-Amino-6-methylpyridazine ≥95%
3-Amino-6-methylpyridazine

Hazard code(s): Irritant

Formula: C₅H₇N₃
MW: 109.13 g/mol
MDL Number: MFCD01569899
CAS Number: 18591-82-7
EINECS: 242-430-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR