3-Methylpyridazine ≥95%

Supplier: Matrix Scientific
Warning

068578-1G 068578-25G 068578-5G
101936-756EA 87.69 USD
101936-756 101936-758 101936-760
3-Methylpyridazine ≥95%
3-Methylpyridazine

Hazard code(s): Irritant

Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 214 °C (1013 hPa)
Density: 1.0491 g/cm³ (20 °C)
MDL Number: MFCD00006469
CAS Number: 1632-76-4
EINECS: 216-640-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR