4-Bromopyridine-2-carbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Bromo-2-cyanopyridine
068399-1G
101935-964EA
21.54
USD
101935-964
4-Bromopyridine-2-carbonitrile ≥95%
4-Bromopyridine-2-carbonitrile
Hazard code(s): Toxic
Formula:
C₆H₃BrN₂ Melting Pt: 87…91 °C |
MDL Number:
MFCD04065805 CAS Number: 62150-45-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...