Methyl-6-bromonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 6-bromonicotinate

068385-1G 068385-5G
101935-902EA 39.1 USD
101935-902 101935-900
Methyl-6-bromonicotinate ≥95%
Methyl 6-bromonicotinate

Hazard code(s): Irritant, Light Sensitive, Stored under Argon

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 106…110 °C
MDL Number: MFCD04972371
CAS Number: 26218-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR