3-Bromo-4-hydroxybenzyl alcohol

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4-(hydroxymethyl)phenol(3-Bromo-4-hydroxyphenyl)methanol

065352-500MG
101929-506EA 364.62 USD
101929-506
3-Bromo-4-hydroxybenzyl alcohol
3-Bromo-4-hydroxybenzyl alcohol
Formula: C₇H₇BrO₂
MW: 203.04 g/mol
MDL Number: MFCD00016869
CAS Number: 29922-56-3
EINECS: 249-957-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR