2-Bromomelatonine

Supplier: Matrix Scientific

Synonyms: N-[2-(2-Bromo-5-methoxy-1H-indol-3-yl)ethyl]-acetamide

065350-500MG
101929-502EA 362.25 USD
101929-502
2-Bromomelatonine
2-Bromomelatonine
Formula: C₁₃H₁₅BrN₂O₂
MDL Number: MFCD01076464
CAS Number: 142959-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR