2',3',4'-Trihydroxyacetophenone

Supplier: Matrix Scientific

Synonyms: Gallacetophenone1-(2,3,4-Trihydroxyphenyl)ethanone

064959-500MG
101928-718EA 146.15 USD
101928-718
2',3',4'-Trihydroxyacetophenone
2',3',4'-Trihydroxyacetophenone
Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 173 °C
MDL Number: MFCD00002193
CAS Number: 528-21-2
EINECS: 208-430-2
Merck Index: 12,04360

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR