1,8-Dinitronaphthalene
Supplier: Matrix Scientific
064579-500MG
101927-964EA
183.08
USD
101927-964
1,8-Dinitronaphthalene
1,8-Dinitronaphthalene
Formula:
C₁₀H₆N₂O₄ MW: 218.17 g/mol Boiling Pt: 390 °C (1013 hPa) Melting Pt: 173 °C |
MDL Number:
MFCD00003918 CAS Number: 602-38-0 EINECS: 210-016-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...