Pentafluorophenylboronic acid

Supplier: Matrix Scientific
Warning

Synonyms: Pentafluorobenzeneboronic acid2,3,4,5,6-Pentafluorobenzeneboronic acidDihydroxy(pentafluorophenyl)borane(Pentafluorophenyl)metaboric acid

064563-500MG
101927-930EA 364.62 USD
101927-930
Pentafluorophenylboronic acid
Pentafluorophenylboronic acid
Formula: C₆H₂BF₅O₂
MW: 211.88 g/mol
MDL Number: MFCD01074663
CAS Number: 1582-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR