4-Ethoxymethylene-2-phenyl-2-oxazoline-5-one

Supplier: Matrix Scientific

Synonyms: 4-Ethoxymethylene-2-phenyloxazolin-5-one(4E)-4-(Ethoxymethylene)-2-phenyl-1,3-oxazol-5(4H)-one

064470-500MG
101927-766EA 183.08 USD
101927-766
4-Ethoxymethylene-2-phenyl-2-oxazoline-5-one
4-Ethoxymethylene-2-phenyl-2-oxazoline-5-one
Formula: C₁₂H₁₁NO₃
MW: 217.22 g/mol
MDL Number: MFCD03225480
CAS Number: 15646-46-5
EINECS: 239-713-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR