2-Ethylcyclopentanone
Supplier: Matrix Scientific
064367-500MG
101927-558EA
183.08
USD
101927-558
2-Ethylcyclopentanone
2-Ethylcyclopentanone
Formula:
C₇H₁₂O MW: 112.17 g/mol |
MDL Number:
MFCD00019314 CAS Number: 4971-18-0 EINECS: 225-612-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...