3-Indolylglyoxylic acid

Supplier: Matrix Scientific

Synonyms: 1H-Indol-3-yl(oxo)acetic acid3-Indoleglyoxylic Acid

063952-500MG
101926-698EA 364.62 USD
101926-698
3-Indolylglyoxylic acid
3-Indolylglyoxylic acid
Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
MDL Number: MFCD00005625
CAS Number: 1477-49-2
EINECS: 216-029-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR