1,6-Dimethylnaphthalene

Supplier: Matrix Scientific

063680-500MG
101926-190EA 189.75 USD
101926-190
1,6-Dimethylnaphthalene
1,6-Dimethylnaphthalene
Formula: C₁₂H₁₂
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: –16.9 °C
Density: 1.009 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004039
CAS Number: 575-43-9
EINECS: 209-385-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR