1-Acenaphthenol

Supplier: Matrix Scientific

Synonyms: 1,2-Dihydroacenaphthylen-1-ol

063665-500MG
101926-160EA 183.08 USD
101926-160
1-Acenaphthenol
1-Acenaphthenol
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
MDL Number: MFCD00003808
CAS Number: 6306-07-6
EINECS: 228-618-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR