5-Bromobenzotriazole

Supplier: Matrix Scientific

Synonyms: 5-Bromo-1H-1,2,3-benzotriazole5-Bromo-1H-benzotriazole

063051-1G
101925-048EA 136.92 USD
101925-048
5-Bromobenzotriazole
5-Bromobenzotriazole
Formula: C₆H₄BrN₃
MW: 198.02 g/mol
MDL Number: MFCD00464510
CAS Number: 32046-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR