4-Quinazolone

Supplier: Matrix Scientific

Synonyms: Quinazolin-4(3H)-one

062975-500MG
101924-914EA 146.15 USD
101924-914
4-Quinazolone
4-Quinazolone
Formula: C₈H₆N₂O
MW: 146.15 g/mol
MDL Number: MFCD00511302
CAS Number: 491-36-1
EINECS: 207-735-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR