N-Aminorhodanine

Supplier: Matrix Scientific

Synonyms: 3-Amino-2-thioxo-1,3-thiazolidin-4-one

062965-500MG
101924-894EA 484.62 USD
101924-894
N-Aminorhodanine
N-Aminorhodanine
Formula: C₃H₄N₂OS₂
MDL Number: MFCD00005489
CAS Number: 1438-16-0
EINECS: 215-870-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR