2-Fluoro-3-pyridineboronic acid pinacol ester

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine2-Fluoropyridine-3-boronic acid pinacol ester

062634-500MG
101924-260EA 243.08 USD
101924-260
2-Fluoro-3-pyridineboronic acid pinacol ester
2-Fluoro-3-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BFNO₂
MW: 223.05 g/mol
MDL Number: MFCD06798245
CAS Number: 452972-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR