2-Chloro-6-fluorocinnamic acid

Supplier: Matrix Scientific

Synonyms: (2E)-3-(2-Chloro-6-fluorophenyl)acrylic acid3-(2-Chloro-6-fluorophenyl)acrylic acid

062321-500MG
101923-734EA 146.15 USD
101923-734
2-Chloro-6-fluorocinnamic acid
2-Chloro-6-fluorocinnamic acid
Formula: C₉H₆ClFO₂
MW: 200.6 g/mol
MDL Number: MFCD00051582
CAS Number: 392-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR