2,6-Dichlorocinnamic acid

Supplier: Matrix Scientific
Warning

Synonyms: (2E)-3-(2,6-Dichlorophenyl)acrylic acid3-(2,6-Dichlorophenyl)acrylic acid

062320-500MG
101923-732EA 146.15 USD
101923-732
2,6-Dichlorocinnamic acid
2,6-Dichlorocinnamic acid
Formula: C₉H₆Cl₂O₂
MW: 217.05 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 192…196 °C
MDL Number: MFCD00004374
CAS Number: 5345-89-1
EINECS: 226-301-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR