4-Quinolinecarboxylic acid
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
Quinoline-4-carboxylic acid
061111-5G
061111-1G
101921-324EA
364.62
USD
101921-324
101921-322
4-Quinolinecarboxylic acid
4-Quinolinecarboxylic acid
Formula:
C₁₀H₇NO₂ MW: 173.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006782 CAS Number: 486-74-8 EINECS: 207-640-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...