5-Nitro-2H-3,1-benzoxazine-2,4(1H)-dione

Supplier: Matrix Scientific

Synonyms: 5-Nitro-1H-benzo[d][1,3]oxazine-2,4-dione

059727-500MG
101918-964EA 306.15 USD
101918-964
5-Nitro-2H-3,1-benzoxazine-2,4(1H)-dione
5-Nitro-2H-3,1-benzoxazine-2,4(1H)-dione
Formula: C₈H₄N₂O₅
MW: 208.13 g/mol
MDL Number: MFCD00016919
CAS Number: 20829-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR