2,5-Dichloro-3-pyridineboronic acid pinacol ester
Supplier: Matrix Scientific
Synonyms:
2,5-Dichloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
, 2,5-Dichloropyridine-3-boronic acid pinacol ester
059477-250MG
101918-232EA
638.46
USD
101918-232
2,5-Dichloro-3-pyridineboronic acid pinacol ester
2,5-Dichloro-3-pyridineboronic acid pinacol ester
Formula:
C₁₁H₁₄BCl₂NO₂ MW: 273.95 g/mol |
MDL Number:
MFCD08063094 CAS Number: 1073371-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...