4-Methoxybenzamidoxime ≥95%

Supplier: Matrix Scientific

Synonyms: N-Hydroxy-4-methoxy-benzamidine

059090-1G
101917-040EA 127.69 USD
101917-040
4-Methoxybenzamidoxime ≥95%
4-Methoxybenzamidoxime

Hazard code(s): Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD00816993
CAS Number: 5373-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR