Methyl-5-fluoronicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-nicotinic acid methyl esterMethyl 5-fluoro-3-pyridinecarboxylate

059070-1G
101916-956EA 146.15 USD
101916-956
Methyl-5-fluoronicotinate ≥95%
Methyl 5-fluoronicotinate

Hazard code(s): Irritant

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
MDL Number: MFCD01692049
CAS Number: 455-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR