2,6-Dichlorobenzaldehyde oxime ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Dichlorobenzaldoxime

058965-1G
101916-516EA 109.85 USD
101916-516
2,6-Dichlorobenzaldehyde oxime ≥95%
2,6-Dichlorobenzaldehyde oxime

Hazard code(s): Irritant

Formula: C₇H₅Cl₂NO
MW: 190.03 g/mol
MDL Number: MFCD00013938
CAS Number: 25185-95-9
EINECS: 246-720-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR