TAPS (3-(Tris(hydroxymethyl)methylamino)propane-1-sulfonic acid)

Supplier: Matrix Scientific

Synonyms: 3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acidN-Tris(hydroxymethyl)methyl-3-aminopropanesulfonic Acid

058420-500MG
101914-774EA 228.85 USD
101914-774
TAPS (3-(Tris(hydroxymethyl)methylamino)propane-1-sulfonic acid)
TAPS (3-(Tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)
Formula: C₇H₁₇NO₆S
MW: 243.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007538
CAS Number: 29915-38-6
EINECS: 249-954-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR