Nitroguanidine stabilized

Supplier: Matrix Scientific

Synonyms: N-Nitroguanidine

058378-500MG
101914-690EA 306.15 USD
101914-690
Nitroguanidine stabilized
Nitroguanidine
Formula: CH₄N₄O₂
MDL Number: MFCD00007039
CAS Number: 556-88-7
EINECS: 209-143-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR