2,4,5-T-butyl ester (2,4,5-trichlorophenoxy acetic acid butyl ester)

Supplier: Matrix Scientific

Synonyms: Butyl (2,4,5-trichlorophenoxy)acetate

058354-500MG
101914-644EA 228.85 USD
101914-644
2,4,5-T-butyl ester (2,4,5-trichlorophenoxy acetic acid butyl ester)
2,4,5-T-butyl ester (2,4,5-trichlorophenoxy acetic acid butyl ester)
Formula: C₁₂H₁₃Cl₃O₃
MW: 311.59 g/mol
MDL Number: MFCD00128016
CAS Number: 93-79-8
EINECS: 202-277-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR