1,8-Dihydroxy-9,10-anthraquinone

Supplier: Matrix Scientific

Synonyms: Dantron1,8-Dihydroxyanthraquinone

058300-500MG
101914-540EA 306.15 USD
101914-540
1,8-Dihydroxy-9,10-anthraquinone
1,8-Dihydroxy-9,10-anthraquinone
Formula: C₁₄H₈O₄
MW: 240.22 g/mol
Melting Pt: 190…193 °C
Density: 1.537 g/cm³ (20 °C)
Flash Pt: >200 °C
MDL Number: MFCD00001211
CAS Number: 117-10-2
EINECS: 204-173-5
Merck Index: 13,02841

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR