Acetylcholine chloride

Supplier: Matrix Scientific

Synonyms: (2-Acetoxyethyl)trimethylammonium chloride2-(Acetyloxy)-N,N,N-trimethylethanaminium chloride

058278-500MG
101914-496EA 15.38 USD
101914-496
Acetylcholine chloride
Acetylcholine chloride
Formula: H₃CC(O)O(CH₂)₂N(CH₃)₃Cl
MW: 181.66 g/mol
Melting Pt: 151 °C
Density: 1.263 g/cm³ (20 °C)
MDL Number: MFCD00011698
CAS Number: 60-31-1
EINECS: 200-468-8
Merck Index: 13,00089

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR