8-Bromo-2-tetralone ≥95%

Supplier: Matrix Scientific

Synonyms: 8-Bromo-3,4-dihydronaphthalen-2(1H)-one

058051-1G
101913-968EA 414 USD
101913-968
8-Bromo-2-tetralone ≥95%
8-Bromo-2-tetralone

Hazard code(s): Irritant

Formula: C₁₀H₉BrO
MDL Number: MFCD03093390
CAS Number: 117294-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR