Thiomorpholine ≥99%

Supplier: Matrix Scientific

057950-5G
101913-786EA 12.65 USD
101913-786
Thiomorpholine ≥99%
Thiomorpholine

Hazard code(s): Corrosive, Air Sensitive

Formula: C₄H₉NS
Boiling Pt: 175…177 °C
Density: 1.086 g/cm³
MDL Number: MFCD00005974
CAS Number: 123-90-0
EINECS: 204-660-2
UN: 3267
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR