1-Benzyl-3-pyrrolidinone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(Phenylmethyl)-3-pyrrolidinone
, 1-Benzylpyrrolidin-3-one
057293-5G
101912-920EA
70.77
USD
101912-920
1-Benzyl-3-pyrrolidinone ≥95%
1-Benzyl-3-pyrrolidinone
Hazard code(s): Irritant, Store in Freezer
Formula:
C₁₁H₁₃NO MW: 175.23 g/mol Boiling Pt: 77…78 °C (0.01mm) Density: 1.091 g/cm³ |
MDL Number:
MFCD00005342 CAS Number: 775-16-6 EINECS: 212-274-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...