(2,4,5-Trimethylphenyl)acetonitrile ≥99%

Supplier: Matrix Scientific

057207-50G
101912-752EA 923.08 USD
101912-752
(2,4,5-Trimethylphenyl)acetonitrile ≥99%
(2,4,5-Trimethylphenyl)acetonitrile

Hazard code(s): Irritant

Formula: C₁₁H₁₃N
Boiling Pt: 95 °C (1mm)
Melting Pt: 29…31 °C
MDL Number: MFCD00060692
CAS Number: 75279-58-2

Order Now

Specification Test Results

Boiling point 95°C/1mm
Melting point 29-31°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR