p-Toluic hydrazide
Supplier: Matrix Scientific
Synonyms:
4-Methylbenzohydrazide
, p-Toluoylhydrazine
, p-Toluic acid hydrazide
, p-Toluic Hydrazide
057193-100G
101912-708EA
458.46
USD
101912-708
p-Toluic hydrazide
p-Toluic hydrazide
Hazard code(s): Irritant
Formula:
C₈H₁₀N₂O MW: 150.18 g/mol Melting Pt: 116…118 °C |
MDL Number:
MFCD00007607 CAS Number: 3619-22-5 EINECS: 222-815-2 |
Specification Test Results
Melting point | 116-118°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...