Phthalamic acid

Supplier: Matrix Scientific

Synonyms: 2-Carbamoylbenzoic acid

057186-10G
101912-682EA 129.95 USD
101912-682
Phthalamic acid
Phthalamic acid

Hazard code(s): Irritant

Formula: C₈H₇NO₃
MW: 165.15 g/mol
Melting Pt: 140…143 °C
MDL Number: MFCD00025476
CAS Number: 88-97-1
EINECS: 201-871-1

Order Now

Specification Test Results

Melting point 140-143°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR